Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Agpat1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163379.1 | NP_001156851.1 | 4 - 5 | 826 | 72 | |
NM_018862.3 | NP_061350.3 | 4 - 5 | 578 | 72 | ||
GenBank mRNA | AB005623.1 | - | 4 - 5 | 824 | 72 | |
AK086281.1 | - | 4 - 5 | 768 | 72 | ||
BC009651.1 | - | 4 - 5 | 578 | 3487719 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |