Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Paxip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_018878.3 | NP_061366.2 | 19 - 20 | 3368 | 73 | |
GenBank mRNA | AF104261.1 | - | 19 - 20 | 3368 | 73 | |
AK144606.1 | - | 19 - 20 | 3230 | 73 | ||
AK148141.1 | - | 19 - 20 | 3241 | 73 | ||
BC066014.1 | - | 19 - 20 | 3157 | 5704566 | 73 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |