Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Exosc9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019393.2 | NP_062266.1 | 6 - 7 | 683 | 85 | |
GenBank mRNA | AF152841.1 | - | 6 - 7 | 656 | 85 | |
AF152842.1 | - | 6 - 7 | 705 | 85 | ||
AK010253.1 | - | 6 - 7 | 658 | 85 | ||
AK011636.1 | - | 6 - 7 | 634 | 85 | ||
AK076096.1 | - | 6 - 7 | 665 | 85 | ||
AK083794.1 | - | 5 - 6 | 922 | 85 | ||
BC005622.1 | - | 6 - 7 | 662 | 3711930 | 85 | |
BC052156.1 | - | 6 - 7 | 655 | 6394529 | 85 | |
BC158075.1 | - | 6 - 7 | 634 | 9007372 | 85 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |