Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nfkb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001177369.1 | NP_001170840.1 | 11 - 12 | 1156 | 76 | |
NM_001177370.1 | NP_001170841.1 | 10 - 11 | 1072 | 76 | ||
NM_019408.3 | NP_062281.1 | 10 - 11 | 1307 | 76 | ||
GenBank mRNA | AF155372.1 | - | 11 - 12 | 1234 | 76 | |
AF155373.1 | - | 11 - 12 | 1271 | 76 | ||
AK134081.1 | - | 11 - 12 | 1575 | 76 | ||
AK137680.1 | - | 11 - 12 | 1146 | 76 | ||
AK144994.1 | - | 11 - 12 | 1149 | 76 | ||
AK145312.1 | - | 10 - 11 | 1273 | 76 | ||
AK148160.1 | - | 11 - 12 | 1144 | 76 | ||
AK150012.1 | - | 11 - 12 | 1135 | 76 | ||
AK153412.1 | - | 6 - 7 | 716 | 76 | ||
BC027423.1 | - | 10 - 11 | 1073 | 3672913 | 76 | |
DQ020182.1 | - | 10 - 11 | 1004 | 76 | ||
DQ020183.1 | - | 9 - 10 | 1125 | 76 | ||
DQ020184.1 | - | 9 - 10 | 1125 | 76 | ||
DQ020185.1 | - | 10 - 11 | 1004 | 76 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Apoptosis Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |