Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nfkb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001177369.1 | NP_001170840.1 | 16 - 17 | 1955 | 67 | |
NM_001177370.1 | NP_001170841.1 | 15 - 16 | 1871 | 67 | ||
NM_019408.3 | NP_062281.1 | 15 - 16 | 2106 | 67 | ||
GenBank mRNA | AF155372.1 | - | 16 - 17 | 2033 | 67 | |
AF155373.1 | - | 16 - 17 | 2070 | 67 | ||
AK134081.1 | - | 16 - 17 | 2374 | 67 | ||
AK137680.1 | - | 16 - 17 | 1945 | 67 | ||
AK144994.1 | - | 16 - 17 | 1948 | 67 | ||
AK145312.1 | - | 15 - 16 | 2072 | 67 | ||
AK148160.1 | - | 16 - 17 | 1943 | 67 | ||
AK150012.1 | - | 16 - 17 | 1934 | 67 | ||
AK153412.1 | - | 11 - 12 | 1515 | 67 | ||
AK196178.1 | - | 1 - 2 | 46 | 67 | ||
BC027423.1 | - | 15 - 16 | 1872 | 3672913 | 67 | |
DQ020182.1 | - | 15 - 16 | 1803 | 67 | ||
DQ020183.1 | - | 14 - 15 | 1924 | 67 | ||
DQ020184.1 | - | 13 - 14 | 2004 | 67 | ||
DQ020185.1 | - | 15 - 16 | 1803 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Apoptosis Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |