Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pus1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025561.3 | NP_001020732.1 | 5 - 6 | 864 | 61 | |
NM_001025562.2 | NP_001020733.1 | 4 - 5 | 507 | 61 | ||
NM_019700.4 | NP_062674.2 | 4 - 5 | 810 | 61 | ||
GenBank mRNA | AB041563.1 | - | 5 - 6 | 865 | 61 | |
AF116237.1 | - | 3 - 4 | 467 | 61 | ||
AK089258.1 | - | 4 - 5 | 810 | 61 | ||
AK138570.1 | - | 2 - 3 | 385 | 61 | ||
AK161521.1 | - | 4 - 5 | 519 | 61 | ||
BC021446.1 | - | 4 - 5 | 686 | 5038201 | 61 | |
BC034359.1 | - | 4 - 5 | 781 | 3595162 | 61 | |
BU515425.1 | - | 4 - 5 | 782 | 61 | ||
CB059120.1 | - | 2 - 3 | 165 | 61 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus Probe spans exons |