Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acpp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019807.2 | NP_062781.2 | 8 - 9 | 928 | 77 | |
NM_207668.2 | NP_997551.1 | 8 - 9 | 928 | 77 | ||
GenBank mRNA | AF210243.1 | - | 8 - 9 | 881 | 77 | |
AK020693.1 | - | 8 - 9 | 890 | 77 | ||
AK029273.1 | - | 8 - 9 | 914 | 77 | ||
AK076383.1 | - | 8 - 9 | 935 | 77 | ||
BC139826.1 | - | 8 - 9 | 919 | 40142642 | 77 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |