Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Noa1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019836.3 | NP_062810.1 | 1 - 2 | 1180 | 69 | |
GenBank mRNA | AB041537.1 | - | 1 - 2 | 1152 | 69 | |
AK011533.1 | - | 1 - 2 | 1172 | 69 | ||
AK017793.1 | - | 1 - 2 | 1153 | 69 | ||
AK019975.1 | - | 1 - 2 | 1162 | 69 | ||
AK128920.1 | - | 1 - 2 | 1152 | 69 | ||
AK139955.1 | - | 1 - 2 | 1124 | 69 | ||
AK157246.1 | - | 1 - 2 | 1180 | 69 | ||
AK163935.1 | - | 1 - 2 | 1154 | 69 | ||
AK172681.1 | - | 1 - 2 | 1131 | 69 | ||
BC005689.1 | - | 1 - 2 | 233 | 69 | ||
BC017154.1 | - | 1 - 2 | 1152 | 4910547 | 69 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus 1700 Inventoried Probe spans exons |