Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pde4b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001177983.1 | NP_001171454.1 | 3 - 4 | 948 | 68 | |
NM_019840.2 | NP_062814.2 | 3 - 4 | 751 | 68 | ||
GenBank mRNA | AF208023.2 | - | 3 - 4 | 686 | 68 | |
AF326555.1 | - | 3 - 4 | 491 | 68 | ||
AF326556.1 | - | 3 - 4 | 491 | 68 | ||
AJ297397.1 | - | 3 - 4 | 686 | 68 | ||
BC049864.1 | - | 3 - 4 | 949 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |