Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Miox | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019977.2 | NP_064361.2 | 4 - 5 | 366 | 95 | |
GenBank mRNA | AF197127.1 | - | 4 - 5 | 364 | 95 | |
AK002406.1 | - | 4 - 5 | 380 | 95 | ||
AY738257.1 | - | 3 - 4 | 336 | 95 | ||
BC013543.1 | - | 4 - 5 | 366 | 4242159 | 95 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |