Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pnkd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039509.1 | NP_001034598.1 | 3 - 4 | 384 | 63 | |
NM_019999.2 | NP_064383.1 | 2 - 3 | 293 | 63 | ||
GenBank mRNA | AB041609.1 | - | 2 - 3 | 294 | 63 | |
AK041238.1 | - | 2 - 3 | 227 | 63 | ||
AK173125.1 | - | 3 - 4 | 358 | 63 | ||
BC058945.1 | - | 4 - 5 | 480 | 6812127 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |