Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem9b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_020050.1 | NP_064434.1 | 3 - 4 | 392 | 84 | |
GenBank mRNA | AK007430.1 | - | 3 - 4 | 360 | 84 | |
AK009147.1 | - | 3 - 4 | 327 | 84 | ||
AK049882.1 | - | 3 - 4 | 479 | 84 | ||
AK077253.1 | - | 3 - 4 | 348 | 84 | ||
AK147127.1 | - | 3 - 4 | 408 | 84 | ||
AK201035.1 | - | 2 - 3 | 206 | 84 | ||
AK211319.1 | - | 2 - 3 | 206 | 84 | ||
BC002208.1 | - | 3 - 4 | 376 | 3490055 | 84 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |