Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0700005887.mm.1 |
TC0700000720.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700005887.mm.1 |
NM_021387.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700005887.mm.1 |
NM_021387.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700005886.mm.1 |
TC0700000720.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700005886.mm.1 |
NM_021387.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700005886.mm.1 |
NM_021387.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700005884.mm.1 |
TC0700000720.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700005884.mm.1 |
NM_021387.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700005884.mm.1 |
NM_021387.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700005885.mm.1 |
TC0700000720.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700005885.mm.1 |
NM_021387.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700005885.mm.1 |
NM_021387.3 |
Refseq |