Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nelfb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021393.2 | NP_067368.2 | 10 - 11 | 1634 | 78 | |
GenBank mRNA | AB041607.1 | - | 9 - 10 | 1755 | 78 | |
AK080419.1 | - | 10 - 11 | 1336 | 78 | ||
AK087524.1 | - | 10 - 11 | 1334 | 78 | ||
AK173124.1 | - | 9 - 10 | 1169 | 78 | ||
BC004762.1 | - | 10 - 11 | 1522 | 3155904 | 78 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Mus musculus Probe spans exons |