Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pga5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021453.4 | NP_067428.2 | 2 - 3 | 311 | 98 | |
GenBank mRNA | AF240776.1 | - | 2 - 3 | 220 | 98 | |
AK004109.1 | - | 2 - 3 | 250 | 98 | ||
AK053965.1 | - | 2 - 3 | 312 | 98 | ||
AK168924.1 | - | 2 - 3 | 250 | 98 | ||
BC119523.2 | - | 2 - 3 | 269 | 40056066 | 98 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Protease Mammalian Gene Collection Mus musculus Probe spans exons |