Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1600000492.mm.1 |
NM_021477.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600000492.mm.1 |
NM_021477.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600000492.mm.1 |
TC1600000054.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1600000482.mm.1 |
NM_021477.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600000482.mm.1 |
NM_021477.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600000482.mm.1 |
TC1600000054.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1600000485.mm.1 |
NM_021477.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600000485.mm.1 |
NM_021477.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600000485.mm.1 |
TC1600000054.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1600000507.mm.1 |
NM_021477.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600000507.mm.1 |
NM_021477.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600000507.mm.1 |
NM_183188.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600000507.mm.1 |
NM_183188.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600000507.mm.1 |
TC1600000054.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1600000483.mm.1 |
NM_021477.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600000483.mm.1 |
NM_021477.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600000483.mm.1 |
TC1600000054.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1600000484.mm.1 |
NM_021477.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600000484.mm.1 |
NM_021477.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600000484.mm.1 |
TC1600000054.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1600000496.mm.1 |
NM_021477.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600000496.mm.1 |
NM_021477.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600000496.mm.1 |
TC1600000054.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1600000500.mm.1 |
NM_183188.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600000500.mm.1 |
NM_183188.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600000500.mm.1 |
TC1600000054.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1600000501.mm.1 |
NM_021477.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600000501.mm.1 |
NM_021477.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600000501.mm.1 |
NM_183188.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600000501.mm.1 |
NM_183188.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600000501.mm.1 |
TC1600000054.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1600000502.mm.1 |
NM_021477.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600000502.mm.1 |
NM_021477.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600000502.mm.1 |
NM_183188.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600000502.mm.1 |
NM_183188.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600000502.mm.1 |
TC1600000054.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1600000508.mm.1 |
NM_021477.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600000508.mm.1 |
NM_021477.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600000508.mm.1 |
NM_183188.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600000508.mm.1 |
NM_183188.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600000508.mm.1 |
TC1600000054.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1600000495.mm.1 |
NM_021477.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600000495.mm.1 |
NM_021477.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600000495.mm.1 |
TC1600000054.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1600000498.mm.1 |
NM_021477.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600000498.mm.1 |
NM_021477.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600000498.mm.1 |
TC1600000054.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1600000503.mm.1 |
NM_021477.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600000503.mm.1 |
NM_021477.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600000503.mm.1 |
NM_183188.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600000503.mm.1 |
NM_183188.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600000503.mm.1 |
TC1600000054.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1600000505.mm.1 |
NM_021477.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600000505.mm.1 |
NM_021477.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600000505.mm.1 |
NM_183188.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600000505.mm.1 |
NM_183188.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600000505.mm.1 |
TC1600000054.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1600000481.mm.1 |
NM_021477.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600000481.mm.1 |
NM_021477.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600000481.mm.1 |
TC1600000054.mm.1 |
MTA TC |