Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pnkp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001290764.1 | NP_001277693.1 | 14 - 15 | 1462 | 65 | |
NM_001290766.1 | NP_001277695.1 | 12 - 13 | 1446 | 65 | ||
NM_001290767.1 | NP_001277696.1 | 12 - 13 | 1217 | 65 | ||
NM_021549.3 | NP_067524.2 | 13 - 14 | 1554 | 65 | ||
GenBank mRNA | AF129451.1 | - | 12 - 13 | 1212 | 65 | |
AK049992.1 | - | 13 - 14 | 1357 | 65 | ||
AK159535.1 | - | 13 - 14 | 1319 | 65 | ||
AK159770.1 | - | 13 - 14 | 1319 | 65 | ||
AK160020.1 | - | 13 - 14 | 1319 | 65 | ||
AK160160.1 | - | 14 - 15 | 1459 | 65 | ||
AK172558.1 | - | 12 - 13 | 1251 | 65 | ||
BC047269.1 | - | 11 - 12 | 1496 | 65 | ||
BC057659.1 | - | 13 - 14 | 1356 | 2865792 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |