Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0400018253.mm.1 |
NM_001177850.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400018253.mm.1 |
NM_001177850.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400018253.mm.1 |
NM_001177852.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400018253.mm.1 |
NM_001177852.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400018253.mm.1 |
NM_001177853.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400018253.mm.1 |
NM_001177853.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400018253.mm.1 |
NM_001177854.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400018253.mm.1 |
NM_001177854.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400018253.mm.1 |
NM_023066.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400018253.mm.1 |
NM_023066.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400018253.mm.1 |
TC0400002189.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400018252.mm.1 |
NM_001177849.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400018252.mm.1 |
NM_001177849.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400018252.mm.1 |
NM_001177850.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400018252.mm.1 |
NM_001177850.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400018252.mm.1 |
NM_001177851.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400018252.mm.1 |
NM_001177851.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400018252.mm.1 |
NM_001177852.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400018252.mm.1 |
NM_001177852.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400018252.mm.1 |
NM_001177853.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400018252.mm.1 |
NM_001177853.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400018252.mm.1 |
NM_001177854.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400018252.mm.1 |
NM_001177854.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400018252.mm.1 |
NM_023066.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400018252.mm.1 |
NM_023066.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400018252.mm.1 |
TC0400002189.mm.1 |
MTA TC |