Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pgpep1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_023217.4 | NP_075706.1 | 3 - 4 | 301 | 100 | |
GenBank mRNA | AJ278829.1 | - | 3 - 4 | 233 | 100 | |
AK003373.1 | - | 3 - 4 | 245 | 100 | ||
AK012658.1 | - | 3 - 4 | 233 | 100 | ||
AK047835.1 | - | 3 - 4 | 245 | 100 | ||
AK049632.1 | - | 3 - 4 | 224 | 100 | ||
AK181842.1 | - | 2 - 3 | 100 | |||
BC043041.1 | - | 3 - 4 | 211 | 100 | ||
BC051938.1 | - | 3 - 4 | 230 | 5717365 | 100 | |
BY313315.1 | - | 3 - 4 | 302 | 100 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Protease Mammalian Gene Collection Mus musculus Probe spans exons |