Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nuf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AF326732.1 | - | 1 - 2 | 108 | 137 | |
AK044882.1 | - | 1 - 2 | 100 | 137 | ||
AK089939.1 | - | 1 - 2 | 93 | 137 | ||
AK162249.1 | - | 1 - 2 | 103 | 137 | ||
BC020026.1 | - | 1 - 2 | 76 | 3587655 | 137 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus 1700 Inventoried Probe spans exons |