Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Plekho1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_023320.2 | NP_075809.1 | 2 - 3 | 213 | 86 | |
GenBank mRNA | AF168675.1 | - | 2 - 3 | 213 | 86 | |
AK012945.1 | - | 2 - 3 | 374 | 86 | ||
AK014374.1 | - | 2 - 3 | 233 | 86 | ||
AK170025.1 | - | 2 - 3 | 328 | 86 | ||
AK170922.1 | - | 2 - 3 | 321 | 86 | ||
BC117003.1 | - | 2 - 3 | 220 | 40126322 | 86 | |
BC125441.1 | - | 2 - 3 | 220 | 40129956 | 86 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |