Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fip1l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001159574.1 | NP_001153046.1 | 11 - 12 | 1193 | 90 | |
GenBank mRNA | AK049672.1 | - | 11 - 12 | 1179 | 90 | |
BC003263.1 | - | 13 - 14 | 1328 | 3499130 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus 1700 Inventoried Probe spans exons |