Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tspan13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025359.3 | NP_079635.1 | 4 - 5 | 588 | 66 | |
GenBank mRNA | AK008175.1 | - | 4 - 5 | 528 | 66 | |
AK012571.1 | - | 4 - 5 | 600 | 66 | ||
AK090217.1 | - | 4 - 5 | 600 | 66 | ||
AK090322.1 | - | 4 - 5 | 600 | 66 | ||
AK158761.1 | - | 4 - 5 | 593 | 66 | ||
AK165768.1 | - | 4 - 5 | 625 | 66 | ||
AK167495.1 | - | 4 - 5 | 640 | 66 | ||
AK167979.1 | - | 4 - 5 | 593 | 66 | ||
BC018317.1 | - | 4 - 5 | 588 | 4166625 | 66 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |