Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dnajc15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025384.3 | NP_079660.1 | 2 - 3 | 172 | 79 | |
GenBank mRNA | AK002598.1 | - | 2 - 3 | 168 | 79 | |
AK003396.1 | - | 2 - 3 | 182 | 79 | ||
AK007806.1 | - | 2 - 3 | 161 | 79 | ||
AK007833.1 | - | 2 - 3 | 161 | 79 | ||
BC027509.1 | - | 2 - 3 | 172 | 1364626 | 79 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |