Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Commd4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025417.2 | NP_079693.1 | 4 - 5 | 209 | 105 | |
GenBank mRNA | AK004174.1 | - | 4 - 5 | 210 | 105 | |
AK009802.1 | - | 4 - 5 | 208 | 105 | ||
AK172194.1 | - | 3 - 4 | 1321 | 105 | ||
AK188693.1 | - | 3 - 4 | 107 | |||
BC033438.1 | - | 3 - 4 | 195 | 3995599 | 105 | |
BC049222.1 | - | 4 - 5 | 208 | 2649359 | 105 | |
BC055798.1 | - | 3 - 4 | 1300 | 105 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |