Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tpd52l2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001291202.1 | NP_001278131.1 | 4 - 5 | 588 | 72 | |
NM_025482.3 | NP_079758.2 | 5 - 6 | 648 | 72 | ||
GenBank mRNA | AK013085.1 | - | 5 - 6 | 590 | 72 | |
AK013188.1 | - | 5 - 6 | 601 | 72 | ||
AK051268.1 | - | 4 - 5 | 532 | 72 | ||
AK159925.1 | - | 5 - 6 | 576 | 72 | ||
BC006886.1 | - | 5 - 6 | 537 | 2647702 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |