Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0300000555.mm.1 |
NM_001163536.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300000555.mm.1 |
NM_001163536.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300000555.mm.1 |
NM_001163537.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300000555.mm.1 |
NM_001163537.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300000555.mm.1 |
NM_025489.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300000555.mm.1 |
NM_025489.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300000555.mm.1 |
TC0300000129.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300000550.mm.1 |
NM_001163536.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300000550.mm.1 |
NM_001163536.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300000550.mm.1 |
NM_025489.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300000550.mm.1 |
NM_025489.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300000550.mm.1 |
TC0300000129.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300000549.mm.1 |
NM_001163536.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300000549.mm.1 |
NM_001163536.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300000549.mm.1 |
NM_025489.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300000549.mm.1 |
NM_025489.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300000549.mm.1 |
TC0300000129.mm.1 |
MTA TC |