Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cutc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113562.1 | NP_001107034.1 | 2 - 3 | 277 | 61 | |
NM_025530.3 | NP_079806.2 | 2 - 3 | 277 | 61 | ||
GenBank mRNA | AK007591.1 | - | 2 - 3 | 202 | 61 | |
AK009702.1 | - | 2 - 3 | 302 | 61 | ||
BC026775.1 | - | 2 - 3 | 187 | 3582902 | 61 | |
BY794464.1 | - | 2 - 3 | 278 | 61 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |