Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aptx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025445.2 | NP_001020616.1 | 3 - 4 | 236 | 82 | |
GenBank mRNA | AK010516.1 | - | 3 - 4 | 223 | 82 | |
AK017522.1 | - | 2 - 3 | 206 | 82 | ||
BC068309.1 | - | 2 - 3 | 149 | 82 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |