Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bpifb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025631.3 | NP_079907.2 | 6 - 7 | 622 | 65 | |
GenBank mRNA | AK009346.1 | - | 6 - 7 | 595 | 65 | |
AK009504.1 | - | 6 - 7 | 597 | 65 | ||
AK009619.1 | - | 6 - 7 | 595 | 65 | ||
AK009754.1 | - | 2 - 3 | 198 | 65 | ||
AK009776.1 | - | 6 - 7 | 579 | 65 | ||
AK009794.1 | - | 6 - 7 | 593 | 65 | ||
AK009871.1 | - | 6 - 7 | 597 | 65 | ||
AK010130.1 | - | 6 - 7 | 579 | 65 | ||
AK010154.1 | - | 6 - 7 | 593 | 65 | ||
AK010157.1 | - | 6 - 7 | 623 | 65 | ||
AK010215.1 | - | 6 - 7 | 593 | 65 | ||
AK028156.1 | - | 6 - 7 | 593 | 65 | ||
AK028158.1 | - | 6 - 7 | 596 | 65 | ||
AK075874.1 | - | 6 - 7 | 593 | 65 | ||
AK160516.1 | - | 6 - 7 | 593 | 65 | ||
BC116681.1 | - | 6 - 7 | 577 | 40106489 | 65 | |
BC116727.1 | - | 6 - 7 | 577 | 40106498 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |