Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bpifb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025631.3 | NP_079907.2 | 14 - 15 | 1375 | 96 | |
GenBank mRNA | AK009346.1 | - | 14 - 15 | 1347 | 96 | |
AK009504.1 | - | 14 - 15 | 1350 | 97 | ||
AK009619.1 | - | 14 - 15 | 1348 | 96 | ||
AK009754.1 | - | 10 - 11 | 951 | 96 | ||
AK009776.1 | - | 14 - 15 | 1333 | 96 | ||
AK009794.1 | - | 14 - 15 | 1335 | 96 | ||
AK009871.1 | - | 14 - 15 | 1344 | 96 | ||
AK010130.1 | - | 14 - 15 | 1330 | 96 | ||
AK010154.1 | - | 14 - 15 | 1343 | 96 | ||
AK010157.1 | - | 14 - 15 | 1374 | 96 | ||
AK010215.1 | - | 14 - 15 | 1344 | 96 | ||
AK028156.1 | - | 14 - 15 | 1346 | 96 | ||
AK028158.1 | - | 14 - 15 | 1349 | 96 | ||
AK160516.1 | - | 14 - 15 | 1346 | 96 | ||
BC116681.1 | - | 14 - 15 | 1330 | 40106489 | 96 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |