Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pspc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025682.3 | NP_079958.3 | 4 - 5 | 1125 | 83 | |
GenBank mRNA | AK011272.1 | - | 4 - 5 | 1122 | 83 | |
AK017689.1 | - | 4 - 5 | 1126 | 83 | ||
AK150823.1 | - | 4 - 5 | 1126 | 83 | ||
AK160722.1 | - | 4 - 5 | 1126 | 83 | ||
BC026772.1 | - | 4 - 5 | 1104 | 3711631 | 83 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |