Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tbc1d15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025706.3 | NP_079982.3 | 4 - 5 | 425 | 88 | |
GenBank mRNA | AK010704.1 | - | 4 - 5 | 357 | 88 | |
AK014477.1 | - | 4 - 5 | 357 | 88 | ||
AK078875.1 | - | 4 - 5 | 371 | 88 | ||
AK147085.1 | - | 4 - 5 | 388 | 88 | ||
BB618866.1 | - | 4 - 5 | 429 | 88 | ||
BC032087.1 | - | 4 - 5 | 353 | 88 | ||
BC053395.1 | - | 4 - 5 | 406 | 30054233 | 88 | |
DQ054831.1 | - | 4 - 5 | 356 | 88 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |