Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Odf2l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001162538.1 | NP_001156010.1 | 8 - 9 | 1058 | 66 | |
NM_001162539.1 | NP_001156011.1 | 7 - 8 | 929 | 66 | ||
NM_025714.4 | NP_079990.2 | 8 - 9 | 924 | 66 | ||
GenBank mRNA | AK014630.1 | - | 8 - 9 | 926 | 66 | |
AK016726.1 | - | 8 - 9 | 1040 | 66 | ||
AK031564.1 | - | 8 - 9 | 1060 | 66 | ||
AK048931.1 | - | 6 - 7 | 2516 | 66 | ||
AK085115.1 | - | 8 - 9 | 1050 | 66 | ||
BC020075.1 | - | 7 - 8 | 898 | 3979963 | 66 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |