Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrrk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025730.3 | NP_080006.3 | 41 - 42 | 6169 | 94 | |
GenBank mRNA | AK014886.1 | - | 2 - 3 | 228 | 94 | |
AK014938.1 | - | 2 - 3 | 241 | 94 | ||
AY792512.1 | - | 41 - 42 | 6112 | 94 | ||
BC034074.1 | - | 7 - 8 | 1052 | 94 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |