Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Proz | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025834.3 | NP_080110.1 | 4 - 5 | 495 | 60 | |
GenBank mRNA | AK005011.2 | - | 4 - 5 | 439 | 60 | |
AK008819.1 | - | 4 - 5 | 496 | 60 | ||
BC024104.1 | - | 4 - 5 | 624 | 5136392 | 60 | |
BC038045.1 | - | 4 - 5 | 460 | 4196827 | 60 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |