Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fank1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025850.2 | NP_080126.1 | 4 - 5 | 468 | 86 | |
GenBank mRNA | AK005694.1 | - | 4 - 5 | 478 | 86 | |
AK017071.1 | - | 4 - 5 | 492 | 86 | ||
AY819025.1 | - | 4 - 5 | 498 | 86 | ||
BC046278.1 | - | 4 - 5 | 468 | 6310911 | 86 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |