Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dhdds | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026144.4 | NP_080420.2 | 4 - 5 | 473 | 72 | |
GenBank mRNA | AK004993.1 | - | 4 - 5 | 535 | 72 | |
AK014317.1 | - | 5 - 6 | 661 | 72 | ||
AK034277.1 | - | 4 - 5 | 535 | 72 | ||
AK036929.1 | - | 4 - 5 | 551 | 72 | ||
AK049991.1 | - | 4 - 5 | 474 | 72 | ||
AK135087.1 | - | 4 - 5 | 535 | 72 | ||
AK149138.1 | - | 4 - 5 | 505 | 72 | ||
AK171207.1 | - | 4 - 5 | 547 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mus musculus Probe spans exons |