Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Car4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007607.2 | NP_031633.1 | 2 - 3 | 244 | 73 | |
GenBank mRNA | AK154754.1 | - | 2 - 3 | 210 | 73 | |
AK165399.1 | - | 2 - 3 | 172 | 73 | ||
BC012704.1 | - | 2 - 3 | 169 | 4217743 | 73 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |