Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cct2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007636.2 | NP_031662.2 | 1 - 2 | 114 | 81 | |
GenBank mRNA | AB041570.1 | - | 1 - 2 | 86 | 81 | |
AK012295.1 | - | 1 - 2 | 70 | 81 | ||
AK075581.1 | - | 1 - 2 | 87 | 81 | ||
AK088145.1 | - | 1 - 2 | 77 | 81 | ||
AK145406.1 | - | 1 - 2 | 115 | 81 | ||
AK166891.1 | - | 1 - 2 | 87 | 81 | ||
BC026918.1 | - | 1 - 2 | 80 | 4500896 | 81 | |
Z31553.1 | - | 1 - 2 | 65 | 81 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus 1700 Inventoried Probe spans exons |