Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cct5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007637.2 | NP_031663.1 | 2 - 3 | 267 | 78 | |
GenBank mRNA | AK077753.1 | - | 2 - 3 | 225 | 78 | |
AK088184.1 | - | 2 - 3 | 201 | 78 | ||
AK129056.1 | - | 2 - 3 | 173 | 78 | ||
AK133857.1 | - | 2 - 3 | 225 | 78 | ||
AK136493.1 | - | 2 - 3 | 219 | 78 | ||
AK145445.1 | - | 2 - 3 | 236 | 78 | ||
AK146405.1 | - | 2 - 3 | 239 | 78 | ||
AK146425.1 | - | 2 - 3 | 219 | 78 | ||
AK150390.1 | - | 2 - 3 | 186 | 78 | ||
AK153911.1 | - | 2 - 3 | 243 | 78 | ||
AK167895.1 | - | 2 - 3 | 208 | 78 | ||
BC092233.1 | - | 2 - 3 | 237 | 78 | ||
BC094427.1 | - | 2 - 3 | 214 | 30537966 | 78 | |
Z31555.1 | - | 2 - 3 | 267 | 78 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Probe spans exons |