Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdk11b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007661.3 | NP_031687.2 | 14 - 15 | 1659 | 100 | |
GenBank mRNA | AK077668.1 | - | 14 - 15 | 1660 | 100 | |
AK147133.1 | - | 14 - 15 | 1637 | 100 | ||
BC025058.1 | - | 3 - 4 | 309 | 100 | ||
BC052920.1 | - | 14 - 15 | 1737 | 6489832 | 100 | |
L37092.1 | - | 13 - 14 | 2231 | 100 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Protein Kinase Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |