Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdh16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001252627.1 | NP_001239556.1 | 15 - 16 | 2448 | 65 | |
NM_001252628.1 | NP_001239557.1 | 16 - 17 | 2538 | 65 | ||
NM_007663.3 | NP_031689.1 | 16 - 17 | 2538 | 65 | ||
GenBank mRNA | AB009292.1 | - | 2 - 3 | 242 | 65 | |
AF016271.1 | - | 16 - 17 | 2406 | 65 | ||
AJ609635.1 | - | 16 - 17 | 2405 | 65 | ||
AK052636.1 | - | 16 - 17 | 2341 | 65 | ||
AK093905.1 | - | 16 - 17 | 2336 | 65 | ||
AK164546.1 | - | 15 - 16 | 2449 | 65 | ||
BC015251.1 | - | 16 - 17 | 2386 | 4235757 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Mus musculus Probe spans exons |