Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Inadl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005787.1 | NP_001005787.1 | 13 - 14 | 1790 | 98 | |
NM_007704.2 | NP_031730.1 | 13 - 14 | 1790 | 98 | ||
NM_172696.2 | NP_766284.2 | 39 - 40 | 5240 | 98 | ||
GenBank mRNA | AF060539.1 | - | 13 - 14 | 1690 | 98 | |
AK077268.1 | - | 26 - 27 | 3522 | 98 | ||
BC037607.1 | - | 13 - 14 | 1462 | 98 | ||
BC057124.1 | - | 13 - 14 | 1469 | 98 | ||
BC082787.1 | - | 28 - 29 | 3633 | 98 | ||
BC141404.1 | - | 39 - 40 | 5216 | 9056017 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |