Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Col1a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007743.2 | NP_031769.2 | 22 - 23 | 1412 | 67 | |
GenBank mRNA | AK132496.1 | - | 22 - 23 | 1408 | 67 | |
AK142111.1 | - | 2 - 3 | 310 | 67 | ||
AK159212.1 | - | 22 - 23 | 1413 | 67 | ||
AK159408.1 | - | 22 - 23 | 1409 | 67 | ||
AK159578.1 | - | 22 - 23 | 1410 | 67 | ||
AK160098.1 | - | 22 - 23 | 1412 | 67 | ||
AK160787.1 | - | 22 - 23 | 1408 | 67 | ||
AK160788.1 | - | 22 - 23 | 1407 | 67 | ||
AK160790.1 | - | 22 - 23 | 1408 | 67 | ||
AK160814.1 | - | 14 - 15 | 877 | 67 | ||
AK160946.1 | - | 22 - 23 | 1411 | 67 | ||
AK161017.1 | - | 22 - 23 | 1413 | 67 | ||
AK161605.1 | - | 22 - 23 | 1408 | 67 | ||
AK164808.1 | - | 22 - 23 | 1408 | 67 | ||
BC007158.1 | - | 22 - 23 | 1392 | 3487477 | 67 | |
BC042503.1 | - | 22 - 23 | 1400 | 3586272 | 67 | |
X58251.1 | - | 22 - 23 | 1348 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |