Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Col1a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007743.2 | NP_031769.2 | 33 - 34 | 2192 | 99 | |
GenBank mRNA | AK075707.1 | - | 11 - 12 | 763 | 99 | |
AK132496.1 | - | 33 - 34 | 2188 | 99 | ||
AK159212.1 | - | 33 - 34 | 2193 | 99 | ||
AK159408.1 | - | 33 - 34 | 2189 | 99 | ||
AK159578.1 | - | 33 - 34 | 2190 | 99 | ||
AK159958.1 | - | 12 - 13 | 811 | 99 | ||
AK160098.1 | - | 33 - 34 | 2192 | 99 | ||
AK160787.1 | - | 33 - 34 | 2188 | 99 | ||
AK160788.1 | - | 33 - 34 | 2187 | 99 | ||
AK160790.1 | - | 33 - 34 | 2188 | 99 | ||
AK160814.1 | - | 25 - 26 | 1657 | 99 | ||
AK160946.1 | - | 33 - 34 | 2191 | 99 | ||
AK161017.1 | - | 33 - 34 | 2193 | 99 | ||
AK161605.1 | - | 33 - 34 | 2188 | 99 | ||
AK164628.1 | - | 12 - 13 | 811 | 99 | ||
AK164808.1 | - | 33 - 34 | 2188 | 99 | ||
BC007158.1 | - | 33 - 34 | 2172 | 3487477 | 99 | |
BC042503.1 | - | 33 - 34 | 2180 | 3586272 | 99 | |
X58251.1 | - | 33 - 34 | 2128 | 99 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |