Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Col1a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007743.2 | NP_031769.2 | 44 - 45 | 3104 | 63 | |
GenBank mRNA | AK075707.1 | - | 22 - 23 | 1676 | 63 | |
AK132496.1 | - | 44 - 45 | 3100 | 63 | ||
AK159120.1 | - | 4 - 5 | 305 | 63 | ||
AK159212.1 | - | 44 - 45 | 3106 | 63 | ||
AK159408.1 | - | 44 - 45 | 3101 | 63 | ||
AK159578.1 | - | 44 - 45 | 3103 | 63 | ||
AK159958.1 | - | 23 - 24 | 1724 | 63 | ||
AK160098.1 | - | 44 - 45 | 3105 | 63 | ||
AK160787.1 | - | 44 - 45 | 3100 | 63 | ||
AK160788.1 | - | 44 - 45 | 3099 | 63 | ||
AK160790.1 | - | 44 - 45 | 3100 | 63 | ||
AK160814.1 | - | 36 - 37 | 2569 | 63 | ||
AK160946.1 | - | 44 - 45 | 3103 | 63 | ||
AK161017.1 | - | 44 - 45 | 3105 | 63 | ||
AK161605.1 | - | 44 - 45 | 3100 | 63 | ||
AK164628.1 | - | 23 - 24 | 1723 | 63 | ||
AK164808.1 | - | 44 - 45 | 3100 | 63 | ||
BC007158.1 | - | 44 - 45 | 3084 | 3487477 | 63 | |
BC012438.1 | - | 5 - 6 | 544 | 63 | ||
BC042503.1 | - | 44 - 45 | 3092 | 3586272 | 63 | |
X58251.1 | - | 44 - 45 | 3040 | 63 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |