Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0800009723.mm.1 |
NM_181322.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800009723.mm.1 |
NM_181322.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800009723.mm.1 |
TC0800001223.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800009714.mm.1 |
NM_181322.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800009714.mm.1 |
NM_181322.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800009714.mm.1 |
TC0800001223.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800009725.mm.1 |
NM_181322.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800009725.mm.1 |
NM_181322.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800009725.mm.1 |
TC0800001223.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800009724.mm.1 |
NM_181322.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800009724.mm.1 |
NM_181322.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800009724.mm.1 |
TC0800001223.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800009721.mm.1 |
NM_181322.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800009721.mm.1 |
NM_181322.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800009721.mm.1 |
TC0800001223.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800009712.mm.1 |
NM_181322.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800009712.mm.1 |
NM_181322.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800009712.mm.1 |
TC0800001223.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800009720.mm.1 |
NM_181322.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800009720.mm.1 |
NM_181322.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800009720.mm.1 |
TC0800001223.mm.1 |
MTA TC |