Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyp4b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007823.2 | NP_031849.1 | 6 - 7 | 785 | 55 | |
GenBank mRNA | AK078882.1 | - | 3 - 4 | 397 | 55 | |
AK143890.1 | - | 6 - 7 | 798 | 55 | ||
AK165445.1 | - | 6 - 7 | 801 | 55 | ||
BC008996.1 | - | 6 - 7 | 789 | 3967680 | 55 | |
D50834.1 | - | 6 - 7 | 785 | 55 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |