Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyp7a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007824.2 | NP_031850.2 | 4 - 5 | 1095 | 111 | |
GenBank mRNA | AK050020.1 | - | 4 - 5 | 1095 | 111 | |
AK050210.1 | - | 4 - 5 | 1096 | 111 | ||
AK050220.1 | - | 4 - 5 | 1095 | 111 | ||
AK050260.1 | - | 4 - 5 | 1095 | 111 | ||
BC130261.1 | - | 4 - 5 | 1065 | 40142789 | 111 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection P450 Mus musculus Mouse Immune Response Inventoried Mouse_Immune_Response Probe spans exons |